Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-tert-butyl-1,3,4-thiadiazole 97%

Supplier: AMBEED, INC
Warning

A407364-10G A407364-1G A407364-25G A407364-5G
77168-360EA 85 USD InStock
77168-360 77168-362 77168-364 77168-366
2-Amino-5-tert-butyl-1,3,4-thiadiazole 97%
2-Amino-5-tert-butyl-1,3,4-thiadiazole
Precaución: for Research Use Only


Formula: C₆H₁₁N₃S
MDL Number: MFCD00040403
CAS Number: 39222-73-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty