Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Betahistine mesylate 99%

Supplier: AMBEED, INC
Warning

A412949-100G A412949-25G A412949-5G
77196-098EA 183 USD InStock
77196-098 77196-100 77196-102
Betahistine mesylate 99%
Betahistine mesylate
Precaución: for Research Use Only


Formula: C₁₀H₂₀N₂O₆S₂
MW: 328.40 g/mol
MDL Number: MFCD00211321
CAS Number: 54856-23-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty