Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 4-aminoisophthalate 95%

Supplier: AMBEED, INC
Warning

A415401-100MG A415401-1G A415401-250MG
77168-466EA 39 USD InStock
77168-466 77168-468 77168-470
Dimethyl 4-aminoisophthalate 95%
Dimethyl 4-aminoisophthalate
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₄
MDL Number: MFCD12674801
CAS Number: 63746-12-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty