Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

11-Mercaptoundecanoic acid 98%

Supplier: AMBEED, INC
Danger

A426627-5G A426627-1G A426627-25G
77196-254EA 76 USD InStock
77196-254 77551-280 77746-859
11-Mercaptoundecanoic acid 98%
11-Mercaptoundecanoic acid
Precaución: for Research Use Only


Formula: C₁₁H₂₂O₂S
MDL Number: MFCD00022096
CAS Number: 71310-21-9
UN: 3335
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty