Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

11-Bromoundecanoic acid 98%

Supplier: AMBEED, INC
Warning

A429977-100G A429977-25G A429977-10G
77196-288EA 6.18 USD InStock
77196-288 77616-850 77616-852 77746-869
11-Bromoundecanoic acid 98%
11-Bromoundecanoic acid
Precaución: for Research Use Only


Formula: BrH₂C(CH₂)₉COOH
MDL Number: MFCD00002732
CAS Number: 2834-05-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty