Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-N-Methyl-1-phenylethanamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A430733-1G A430733-250MG
77168-644EA 41 USD InStock
77168-644 77168-646
(R)-N-Methyl-1-phenylethanamine hydrochloride 95%
(R)-N-Methyl-1-phenylethanamine hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₄ClN
MDL Number: MFCD28138033
CAS Number: 1096105-18-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty