Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxycarbonyl)piperazine-2-carboxylic acid 95%

Supplier: AMBEED, INC
Danger

A432600-100MG A432600-1G A432600-250MG A432600-5G
77168-670EA 72 USD InStock
77168-670 77168-672 77168-674 77168-676
(R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxycarbonyl)piperazine-2-carboxylic acid 95%
(R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxycarbonyl)piperazine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₂₅H₂₈N₂O₆
MDL Number: MFCD02179100
CAS Number: 1217723-28-8
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty