Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl-4-oxoazepane-1-carboxylate 98%

Supplier: AMBEED, INC
Warning

A433166-10G A433166-1G A433166-250MG A433166-25G A433166-5G
77168-688EA 139 USD InStock
77168-688 77168-690 77168-692 77168-694 77168-696
Benzyl-4-oxoazepane-1-carboxylate 98%
Benzyl-4-oxoazepane-1-carboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₇NO₃
MDL Number: MFCD03788442
CAS Number: 83621-33-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty