Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,4,5-Tetramethoxybenzene 97%

Supplier: AMBEED, INC
Warning

A438934-100MG A438934-1G A438934-250MG
77196-348EA 96 USD InStock
77196-348 77196-350 77196-352
1,2,4,5-Tetramethoxybenzene 97%
1,2,4,5-Tetramethoxybenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₄O₄
MDL Number: MFCD00094450
CAS Number: 2441-46-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty