Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Fluorophenyl)-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: AMBEED, INC
Warning

A440986-100MG A440986-1G A440986-250MG
77196-360EA 70 USD InStock
77196-360 77196-362 77196-364
1-(2-Fluorophenyl)-1,2,3,4-tetrahydroisoquinoline 97%
1-(2-Fluorophenyl)-1,2,3,4-tetrahydroisoquinoline
Precaución: for Research Use Only


Formula: C₁₅H₁₄FN
MDL Number: MFCD02179249
CAS Number: 1017125-85-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty