Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-N,N-diisopropylnicotinamide 95+%

Supplier: AMBEED, INC
Warning

A441914-100MG A441914-10G A441914-1G A441914-250MG A441914-5G
77168-800EA 5 USD InStock
77168-800 77168-802 77168-804 77168-806 77168-808
6-Chloro-N,N-diisopropylnicotinamide 95+%
6-Chloro-N,N-diisopropylnicotinamide
Precaución: for Research Use Only


Formula: C₁₂H₁₇ClN₂O
MW: 240.72 g/mol
MDL Number: MFCD26127469
CAS Number: 905273-87-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty