Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Spiro[5.5]undecan-2-one 95%

Supplier: AMBEED, INC
Warning

A447965-100MG A447965-250MG A447965-25MG A447965-50MG
77168-854EA 61 USD InStock
77168-854 77168-856 77168-858 77168-860
Spiro[5.5]undecan-2-one 95%
Spiro[5.5]undecan-2-one
Precaución: for Research Use Only


Formula: C₁₁H₁₈O
MDL Number: MFCD00090150
CAS Number: 1781-81-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty