Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carmustine 97%

Supplier: AMBEED, INC
Danger

A448630-10MG A448630-25MG A448630-50MG
77196-456EA 5 USD InStock
77196-456 77196-458 77196-460
Carmustine 97%
Carmustine
Precaución: for Research Use Only


Formula: C₅H₉Cl₂N₃O₂
MDL Number: MFCD00057706
CAS Number: 154-93-8
UN: 2811
ADR: 6.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty