Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dirhodium(II) tetrakis (caprolactam) 98%

Supplier: AMBEED, INC
Warning

A450458-100MG A450458-250MG
77168-874EA 87 USD InStock
77168-874 77168-876
Dirhodium(II) tetrakis (caprolactam) 98%
Dirhodium(II) tetrakis (caprolactam)
Precaución: for Research Use Only


Formula: C₂₄H₄₀N₄O₄Rh₂
MW: 654.419 g/mol
MDL Number: MFCD00467690
CAS Number: 138984-26-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty