Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Di-tert-butylbenzene 95%

Supplier: AMBEED, INC
Danger

A456719-1G A456719-25G A456719-5G
77196-536EA 11 USD InStock
77196-536 77196-538 77196-540
1,4-Di-tert-butylbenzene 95%
1,4-Di-tert-butylbenzene
Precaución: for Research Use Only


Formula: C₁₄H₂₂
MW: 190.33 g/mol
Boiling Pt: 236
MDL Number: MFCD00008836
CAS Number: 1012-72-2
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty