Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methylsulfonyl)piperidine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A457079-100MG A457079-1G A457079-250MG
77196-544EA 88 USD InStock
77196-544 77196-546 77196-548
3-(Methylsulfonyl)piperidine hydrochloride 97%
3-(Methylsulfonyl)piperidine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₄ClNO₂S
MW: 199.7 g/mol
MDL Number: MFCD17015904
CAS Number: 1378304-65-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty