Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-(trifluoromethyl)benzoic acid 97%

Supplier: AMBEED, INC
Warning

A457362-100G A457362-1G A457362-250MG A457362-25G A457362-5G
77196-550EA 245 USD InStock
77196-550 77196-552 77196-554 77196-556 77196-558 77196-560
2-Chloro-3-(trifluoromethyl)benzoic acid 97%
2-Chloro-3-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₄ClF₃O₂
MW: 224.57
MDL Number: MFCD00792415
CAS Number: 39226-97-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty