Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride 97%

Supplier: AMBEED, INC
Danger

A459145-10G A459145-25G A459145-5G
77196-584EA 6 USD InStock
77196-584 77196-586 77196-588
2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride 97%
2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₀ClNO₂
MW: 224.08 g/mol
MDL Number: MFCD02181083
CAS Number: 72830-09-2
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty