Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1-(Dimethylamino)ethyl)phenyl dimethylcarbamate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A467676-100MG A467676-250MG
77169-022EA 44 USD InStock
77169-022 77169-024
3-(1-(Dimethylamino)ethyl)phenyl dimethylcarbamate hydrochloride 98%
3-(1-(Dimethylamino)ethyl)phenyl dimethylcarbamate hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₂₁ClN₂O₂
MDL Number: MFCD28400997
CAS Number: 105601-13-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty