Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromonicotinaldehyde 97%

Supplier: AMBEED, INC
Danger

A467679-10G A467679-25G
77169-026EA 104 USD InStock
77169-026 77169-028
2,4-Dibromonicotinaldehyde 97%
2,4-Dibromonicotinaldehyde
Precaución: for Research Use Only


Formula: C₆H₃Br₂NO
MW: 264.9 g/mol
MDL Number: MFCD10574708
CAS Number: 128071-91-0
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty