Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-aminopentanoate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A480659-1G A480659-250MG
77196-792EA 18 USD InStock
77196-792 77196-794
Methyl 2-aminopentanoate hydrochloride 95%
Methyl 2-aminopentanoate hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₃NO₂·ClH
MDL Number: MFCD01632063
CAS Number: 10047-10-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty