Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyclopropyl-2,2,2-trifluoroethanamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A481712-100MG A481712-1G A481712-250MG
77169-154EA 83 USD InStock
77169-154 77169-156 77169-158
1-Cyclopropyl-2,2,2-trifluoroethanamine hydrochloride 98%
1-Cyclopropyl-2,2,2-trifluoroethanamine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₉ClF₃N
MDL Number: MFCD23098781
CAS Number: 75702-99-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty