Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Pyridyl)-D-alanine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A481822-10G A481822-1G A481822-5G
77196-814EA 760 USD InStock
77196-814 77196-816 77196-818
3-(3-Pyridyl)-D-alanine hydrochloride 95%
3-(3-Pyridyl)-D-alanine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O₂·ClH
MDL Number: MFCD03095567
CAS Number: 350228-35-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty