Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl 1-Boc-piperidine-3-carboxylate 95%

Supplier: AMBEED, INC
Warning

A489883-100MG A489883-1G A489883-250MG A489883-5G
77169-220EA 5 USD InStock
77169-220 77169-222 77169-224 77169-226
Benzyl 1-Boc-piperidine-3-carboxylate 95%
Benzyl 1-Boc-piperidine-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₈H₂₅NO₄
MW: 319.4 g/mol
MDL Number: MFCD11975554
CAS Number: 139985-95-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty