Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5'-phenyl-1,1':3',1''-terphenyl 97%

Supplier: AMBEED, INC
Warning

A491976-100G A491976-250MG A491976-5G
77169-258EA 845 USD InStock
77169-258 77169-260 77169-262
3-Bromo-5'-phenyl-1,1':3',1''-terphenyl 97%
3-Bromo-5'-phenyl-1,1':3',1''-terphenyl
Precaución: for Research Use Only


Formula: C₂₄H₁₇Br
MDL Number: MFCD28134530
CAS Number: 1233200-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty