Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,4,6,7-Tetrahydropyrano[4,3-c]pyrazol-3-yl)methanamine 97%

Supplier: AMBEED, INC
Warning

A501185-100MG A501185-1G
77197-020EA 229 USD InStock
77197-020 77197-022
(2,4,6,7-Tetrahydropyrano[4,3-c]pyrazol-3-yl)methanamine 97%
(2,4,6,7-Tetrahydropyrano[4,3-c]pyrazol-3-yl)methanamine
Precaución: for Research Use Only


Formula: C₇H₁₁N₃O
MDL Number: MFCD20491784
CAS Number: 933722-15-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty