Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α'-Bis(di-t-butylphosphino)-o-xylene 98%

Supplier: AMBEED, INC
Warning

A503246-100MG A503246-1G A503246-250MG A503246-5G
77197-044EA 20 USD InStock
77197-044 77197-046 77197-048 77197-050
α,α'-Bis(di-t-butylphosphino)-o-xylene 98%
α,α'-Bis(di-t-butylphosphino)-o-xylene
Precaución: for Research Use Only


Formula: C₂₄H₄₄P₂
MW: 394.56 g/mol
MDL Number: MFCD03094573
CAS Number: 121954-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty