Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

FH535 ≥98%

Supplier: AMBEED, INC
Danger

A504800-10MG A504800-1MG
77197-070EA 50 USD InStock
77197-070 77197-072
FH535 ≥98%
FH535
Precaución: for Research Use Only


Formula: C₁₃H₁₀Cl₂N₂O₄S
MDL Number: MFCD01212888
CAS Number: 108409-83-2
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty