Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-(aminomethyl)benzoate 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A512527-1G A512527-5G
77169-432EA 82 USD InStock
77169-432 77684-335
tert-Butyl 4-(aminomethyl)benzoate 97%, Ambeed.Inc
tert-Butyl 4-(aminomethyl)benzoate
Precaución: For research use only.


Formula: C₁₂H₁₇NO₂
MDL Number: MFCD04973451
CAS Number: 107045-28-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty