Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Fluoro-4-nitrophenyl)thiomorpholine 97%

Supplier: AMBEED, INC
Warning

A514357-1G A514357-250MG A514357-25G A514357-5G
77197-164EA 11 USD InStock
77197-164 77197-166 77197-168 77197-170
4-(2-Fluoro-4-nitrophenyl)thiomorpholine 97%
4-(2-Fluoro-4-nitrophenyl)thiomorpholine
Precaución: for Research Use Only


Formula: C₁₀H₁₁FN₂O₂S
MDL Number: MFCD11039549
CAS Number: 168828-70-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty