Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,5-Trimethyladamantane 95%

Supplier: AMBEED, INC
Warning

A515605-100MG A515605-1G A515605-250MG A515605-25G A515605-5G
77197-190EA 19 USD InStock
77197-190 77197-192 77197-194 77197-196 77197-198
1,3,5-Trimethyladamantane 95%
1,3,5-Trimethyladamantane
Precaución: for Research Use Only


Formula: C₁₃H₂₂
MDL Number: MFCD00216219
CAS Number: 707-35-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty