Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,9-Decadiene 98%

Supplier: AMBEED, INC
Danger

A515748-25G A515748-5G
77197-206EA 104 USD InStock
77197-206 77197-208
1,9-Decadiene 98%
1,9-Decadiene
Precaución: for Research Use Only


Formula: C₁₀H₁₈
MW: 138.25 g/mol
Boiling Pt: 169
Melting Pt: -71.8
MDL Number: MFCD00008670
CAS Number: 1647-16-1
UN: 3295
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty