Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Fluoro-2-nitrophenyl)piperidine 99%

Supplier: AMBEED, INC
Warning

A524779-1G A524779-25G A524779-5G
77197-282EA 9 USD InStock
77197-282 77197-284 77197-286
1-(4-Fluoro-2-nitrophenyl)piperidine 99%
1-(4-Fluoro-2-nitrophenyl)piperidine
Precaución: for Research Use Only


Formula: C₁₁H₁₃FN₂O₂
MDL Number: MFCD00798370
CAS Number: 719-70-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty