Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PD 169316 95%

Supplier: AMBEED, INC
Danger

A530681-10MG A530681-1MG A530681-25MG A530681-5MG
77169-584EA 33 USD InStock
77169-584 77169-586 77169-588 77169-590
PD 169316 95%
PD 169316
Precaución: for Research Use Only


Formula: C₂₀H₁₃FN₄O₂
MW: 360.34 g/mol
MDL Number: MFCD12405019
CAS Number: 152121-53-4
UN: 2923
ADR: 8,6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty