Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-2-methoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A541123-10G A541123-1G A541123-250MG A541123-5G A541123-25G
77197-392EA 37 USD InStock
77197-392 77197-394 77197-396 77197-398 77747-107
4-Fluoro-2-methoxyphenylboronic acid 98%
4-Fluoro-2-methoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD03788423
CAS Number: 179899-07-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty