Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%

Supplier: AMBEED, INC
Danger

A545420-100MG A545420-1G A545420-250MG
77197-454EA 30 USD InStock
77197-454 77197-456 77197-458
(R)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%
(R)-3,3'-Bis(4-nitrophenyl)-[1,1'-binaphthalene]-2,2'-diol
Precaución: for Research Use Only


Formula: C₃₂H₂₀N₂O₆
MW: 528.511 g/mol
MDL Number: MFCD28139323
CAS Number: 791616-60-9
UN: 1325
ADR: 4.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty