Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((4-Fluorophenyl)ethynyl)aniline 97%

Supplier: AMBEED, INC
Warning

A551315-100MG A551315-1G A551315-250MG
77197-490EA 18 USD InStock
77197-490 77197-492 77197-494
2-((4-Fluorophenyl)ethynyl)aniline 97%
2-((4-Fluorophenyl)ethynyl)aniline
Precaución: for Research Use Only


Formula: C₁₄H₁₀FN
MDL Number: MFCD00168851
CAS Number: 1173153-20-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty