Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

EPZ005687 98+%

Supplier: AMBEED, INC
Warning

A553761-10MG A553761-1MG A553761-25MG A553761-5MG
77197-528EA 44 USD InStock
77197-528 77197-530 77197-532 77197-534
EPZ005687 98+%
EPZ005687
Precaución: for Research Use Only


Formula: C₃₂H₃₇N₅O₃
MDL Number: MFCD25372029
CAS Number: 1396772-26-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty