Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Bromo-3,4-difluoroacetophenone 97%

Supplier: AMBEED, INC
Danger

A553771-1G A553771-5G
77197-536EA 18 USD InStock
77197-536 77747-140
ω-Bromo-3,4-difluoroacetophenone 97%
ω-Bromo-3,4-difluoroacetophenone
Precaución: for Research Use Only


Formula: C₈H₅BrF₂O
MW: 235.03
MDL Number: MFCD00085010
CAS Number: 40706-98-7
UN: 3265
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty