Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-cyclopropylacetate 95%

Supplier: AMBEED, INC
Danger

A554363-250MG A554363-5G
77169-774EA 18 USD InStock
77169-774 77169-776
Ethyl 2-cyclopropylacetate 95%
Ethyl 2-cyclopropylacetate
Precaución: for Research Use Only


Formula: C₇H₁₂O₂
MDL Number: MFCD11976041
CAS Number: 53432-87-4
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty