Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-8-chloro-1,5-naphthyridine 95%

Supplier: AMBEED, INC
Warning

A556450-100MG A556450-1G A556450-250MG
77169-780EA 33 USD InStock
77169-780 77169-782 77169-784
3-Bromo-8-chloro-1,5-naphthyridine 95%
3-Bromo-8-chloro-1,5-naphthyridine
Precaución: for Research Use Only


Formula: C₈H₄BrClN₂
MW: 243.48 g/mol
MDL Number: MFCD11848377
CAS Number: 97267-61-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty