Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-(chloromethyl)pyridine hydrochloride 95%

Supplier: AMBEED, INC
Danger

A559574-100MG A559574-1G A559574-250MG
77169-810EA 39 USD InStock
77169-810 77169-812 77169-814
3-Chloro-5-(chloromethyl)pyridine hydrochloride 95%
3-Chloro-5-(chloromethyl)pyridine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₅Cl₂N·ClH
MW: 198.478 g/mol
MDL Number: MFCD16037340
CAS Number: 847610-86-0
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty