Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-(methoxycarbonyl)benzeneboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A563563-100MG A563563-1G A563563-250MG A563563-5G
77169-834EA 22 USD InStock
77169-834 77169-836 77169-838 77169-840
3-Bromo-5-(methoxycarbonyl)benzeneboronic acid pinacol ester 98%
3-Bromo-5-(methoxycarbonyl)benzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₈BBrO₄
MDL Number: MFCD12405334
CAS Number: 1025718-78-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty