Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diaminonicotinonitrile 97%

Supplier: AMBEED, INC
Warning

A572391-250MG A572391-25MG
77197-750EA 94 USD InStock
77197-750 77197-752
2,6-Diaminonicotinonitrile 97%
2,6-Diaminonicotinonitrile
Precaución: for Research Use Only


Formula: C₆H₆N₄
MW: 134.13 g/mol
MDL Number: MFCD18261970
CAS Number: 99761-27-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty