Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-3,5-dichlorobenzoate 97%

Supplier: AMBEED, INC
Warning

A572584-1G A572584-250MG A572584-5G
77197-762EA 29 USD InStock
77197-762 77197-764 77197-766
Methyl 4-bromo-3,5-dichlorobenzoate 97%
Methyl 4-bromo-3,5-dichlorobenzoate
Precaución: for Research Use Only


Formula: C₈H₅BrCl₂O₂
MW: 283.93 g/mol
MDL Number: MFCD14704248
CAS Number: 117738-81-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty