Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxyresorcinol 98%

Supplier: AMBEED, INC
Warning

A575632-1G A575632-250MG
77197-778EA 39 USD InStock
77197-778 77197-780
5-Methoxyresorcinol 98%
5-Methoxyresorcinol
Precaución: for Research Use Only


Formula: C₇H₈O₃
MDL Number: MFCD00002285
CAS Number: 2174-64-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty