Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethylaniline hydrochloride 98%

Supplier: AMBEED, INC
Warning

A588250-1G A588250-25G A588250-5G
77197-848EA 21 USD InStock
77197-848 77197-850 77197-852
2,3-Dimethylaniline hydrochloride 98%
2,3-Dimethylaniline hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₁N
MDL Number: MFCD00054332
CAS Number: 5417-45-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty