Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyclopropanecarbonyl-2,3-dihydro-1H-indole-5-sulfonyl chloride 97%

Supplier: AMBEED, INC
Danger

A588562-100MG A588562-250MG A588562-5G
77197-854EA 43 USD InStock
77197-854 77197-856 77197-858
1-Cyclopropanecarbonyl-2,3-dihydro-1H-indole-5-sulfonyl chloride 97%
1-Cyclopropanecarbonyl-2,3-dihydro-1H-indole-5-sulfonyl chloride
Precaución: for Research Use Only


Formula: C₁₂H₁₂ClNO₃S
MDL Number: MFCD08687522
CAS Number: 879562-21-7
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty