Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-chloro-2-fluorobenzoic acid 98%

Supplier: AMBEED, INC
Warning

A590951-100MG A590951-1G A590951-250MG A590951-5G
77170-040EA 18 USD InStock
77170-040 77170-042 77170-044 77170-046
4-Bromo-3-chloro-2-fluorobenzoic acid 98%
4-Bromo-3-chloro-2-fluorobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃BrClFO₂
MDL Number: MFCD16622238
CAS Number: 194804-94-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty