Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Propionylfuran 98+%

Supplier: AMBEED, INC
Warning

A596246-1G A596246-250MG A596246-25G A596246-5G
77197-946EA 44 USD InStock
77197-946 77197-948 77197-950 77197-952
2-Propionylfuran 98+%
2-Propionylfuran
Precaución: for Research Use Only


Formula: C₇H₈O₂
MDL Number: MFCD00020936
CAS Number: 3194-15-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty